Learn More
Sigma Aldrich Fine Chemicals Biosciences Poly-L-ornithine hydrobromide, 27378-49-0, MFCD00237231, 50 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P365550MG
Encompass
Mol wt 30,000-70,000. Poly-L-ornithine hydrobromide has been used to coat coverslips in culture vessels for culturing neural progenitor cells. It has also been used to coat culture plates for culturing cerebral cells of mice.
Catalog No. NC1573512