Learn More
Medchemexpress LLC Poly-L-ornithine hydrobromide | 27378-49-0 | 30000-70000 | 25 MG
SDP
Catalog No. 5000438745
Encompass_Preferred
Poly-L-ornithine hydrobromide is a poly-lysine derivative with a molecular weight ranging from 30000 to 70000. It binds to the surface of cell culture vessels through positively charged amino acid residues, forming a coating that promotes cell adhesion and provides cells with a matrix environment required for growth. It is used as a coating agent in cell culture and for the study of primary culture of neurons. It is identified for use as a laboratory chemical.
  • Used as a coating agent in cell culture
  • Promotes cell adhesion
  • Provides cells with a matrix environment required for growth
  • Supports the study of primary culture of neurons, including dopaminergic neurons and oligodendrocytes
  • Can serve as a first layer matrix for cell culture plates
  • Provides a stable adhesion surface for differentiation study of neural stem cells
Catalog No. 50-004-38745 Supplier Medchemexpress LLC Supplier No. 25MGHY126436C
Poly-L-ornithine hydrobromide is a poly-lysine derivative with a molecular weight ranging from 30000 to 70000. It binds to the surface of cell culture vessels through positively charged amino acid residues, forming a coating that promotes cell adhesion and provides cells with a matrix environment required for growth. It is used as a coating agent in cell culture and for the study of primary culture of neurons. It is identified for use as a laboratory chemical.
  • Used as a coating agent in cell culture
  • Promotes cell adhesion
  • Provides cells with a matrix environment required for growth
  • Supports the study of primary culture of neurons, including dopaminergic neurons and oligodendrocytes
  • Can serve as a first layer matrix for cell culture plates
  • Provides a stable adhesion surface for differentiation study of neural stem cells