Learn More
Medchemexpress LLC Poly-L-ornithine hydrobromide | 27378-49-0 | 30000-70000 | 25 MG
SDP
Catalog No. 5000439093
Encompass_Preferred
Poly-L-ornithine hydrobromide is a poly-lysine derivative that binds to the surface of cell culture vessels through positively charged amino acid residues. It forms a coating that promotes cell adhesion and provides a matrix environment required for cell growth. It is used as a coating agent in cell culture and for the study of primary culture of neurons.
  • Poly-lysine derivative.
  • Binds to the surface of cell culture vessels through positively charged amino acid residues.
  • Forms a coating that promotes cell adhesion.
  • Provides a matrix environment required for growth.
  • Used as a coating agent in cell culture.
  • Can be used for the study of primary culture of neurons.
  • Provides a stable and reliable adhesion surface for the differentiation study of neural stem cells.
Catalog No. 50-004-39093 Supplier Medchemexpress LLC Supplier No. 25MGHY126436A
Poly-L-ornithine hydrobromide is a poly-lysine derivative that binds to the surface of cell culture vessels through positively charged amino acid residues. It forms a coating that promotes cell adhesion and provides a matrix environment required for cell growth. It is used as a coating agent in cell culture and for the study of primary culture of neurons.
  • Poly-lysine derivative.
  • Binds to the surface of cell culture vessels through positively charged amino acid residues.
  • Forms a coating that promotes cell adhesion.
  • Provides a matrix environment required for growth.
  • Used as a coating agent in cell culture.
  • Can be used for the study of primary culture of neurons.
  • Provides a stable and reliable adhesion surface for the differentiation study of neural stem cells.