Learn More
Medchemexpress LLC Poly-L-ornithine hydrobromide | 27378-49-0 | MFCD00237231 | 5,000-15,000 | C15H33Br3N6O3X2 | 5 MG
SDP
Catalog No. 5000264382
Encompass_Preferred
Poly-L-ornithine (hydrobromide) is a positively charged polypeptide supplied as a white to off-white solid. The hydrobromide salt with an average molecular weight of 5,000-15,000 is commonly used as a substrate/coating reagent to promote cell adhesion in in vitro cell culture applications. Sold for research use only; consult the SDS and batch Certificate of Analysis for lot-specific details.
  • High molecular weight polymer (5,000-15,000) suitable for cell adhesion coatings.
  • Hydrobromide salt form, soluble in aqueous buffers.
  • Supplied as a white to off-white solid for laboratory handling.
  • Intended for research use only; not for clinical use.
  • Available in small laboratory pack sizes to support assay development.
Catalog No. 50-002-64382 Supplier Medchemexpress LLC Supplier No. HY126436C5MG
Poly-L-ornithine (hydrobromide) is a positively charged polypeptide supplied as a white to off-white solid. The hydrobromide salt with an average molecular weight of 5,000-15,000 is commonly used as a substrate/coating reagent to promote cell adhesion in in vitro cell culture applications. Sold for research use only; consult the SDS and batch Certificate of Analysis for lot-specific details.
  • High molecular weight polymer (5,000-15,000) suitable for cell adhesion coatings.
  • Hydrobromide salt form, soluble in aqueous buffers.
  • Supplied as a white to off-white solid for laboratory handling.
  • Intended for research use only; not for clinical use.
  • Available in small laboratory pack sizes to support assay development.