Learn More
Medchemexpress LLC Poly-L-ornithine hydrochloride | 26982-21-8 | 15,000-30,000 | (C5H12N2O2)n · xHCl | 5 MG
SDP
Catalog No. 5000263885
Encompass_Preferred
Poly-L-ornithine hydrochloride (MW 15,000-30,000) is a polycationic homopolymer of L-ornithine used in research applications to adsorb negatively charged species and to form polyelectrolyte complexes and core-shell carriers for controlled delivery of growth factors. It is commonly used in studies of diabetes and bone regeneration and is supplied as a solid for laboratory use.
  • Polycationic homopolymer with MW 15,000-30,000.
  • Solid, white to off-white appearance.
  • Adsorbs negatively charged species via electrostatic interactions.
  • Enables construction of polyelectrolyte complexes and core-shell carriers for controlled delivery of growth factors.
  • Available in small research-scale package sizes.
Catalog No. 50-002-63885 Supplier Medchemexpress LLC Supplier No. HY126436D5MG
Poly-L-ornithine hydrochloride (MW 15,000-30,000) is a polycationic homopolymer of L-ornithine used in research applications to adsorb negatively charged species and to form polyelectrolyte complexes and core-shell carriers for controlled delivery of growth factors. It is commonly used in studies of diabetes and bone regeneration and is supplied as a solid for laboratory use.
  • Polycationic homopolymer with MW 15,000-30,000.
  • Solid, white to off-white appearance.
  • Adsorbs negatively charged species via electrostatic interactions.
  • Enables construction of polyelectrolyte complexes and core-shell carriers for controlled delivery of growth factors.
  • Available in small research-scale package sizes.