Learn More
Sigma Aldrich Fine Chemicals Biosciences Poly-L-ornithine solution, 27378-49-0, MFCD00286305, 50mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P495750ML
Encompass
Mol Wt 30,000-70,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture. Poly-L-ornithine is a positively charged synthetic amino acid polymer.
Catalog No. 50-165-6954