Learn More
Akina Inc Poly(lactic-co-glycolic acid)-b-Poly(ethylene glycol)-b-Poly(lactic-co-glycolic acid) copolymers (per g)
SDP
Supplier:  Akina Inc AK1841G
Encompass
PLGA-PEG-PLGA (4000-1500-4000 Da) Poly(lactic-co-glycolic acid)-b-Poly(ethylene glycol)-b-Poly(lactic-co-glycolic acid) copolymers (per g)
Catalog No. NC2855779