Learn More
Akina Inc Poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AP0371G
Encompass
PLGA LG 50:50 acid endcap (Mn 1,000-5,000 Da) Poly(lactide-co-glycolide) (per g)
Catalog No. NC1522298