Learn More
Akina Inc Poly(lactide-co-glycolide) (per g)
SDP
Catalog No. NC1576969
Encompass
PLGA LG 50:50 acid endcap (Mn 35,000-45,000 Da) Poly(lactide-co-glycolide) (per g)
Catalog No. NC1576969 Supplier Akina Inc Supplier No. AP0451G
PLGA LG 50:50 acid endcap (Mn 35,000-45,000 Da) Poly(lactide-co-glycolide) (per g)