Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Catalog No. NC2229290
Encompass
PLGA-PEG-PLGA (Mw ~5,000:1,000:5,000 Da) LA:GA 90:10 Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
This item is not returnable. View return policy
Catalog No. NC2229290 Supplier Akina Inc Supplier No. AK0641G
PLGA-PEG-PLGA (Mw ~5,000:1,000:5,000 Da) LA:GA 90:10 Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)