Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Catalog No. NC3713324
Encompass
PLGA-PEG-PLGA (Mw ~1,750-1,500-1,750 Da) LA:GA 15:1 Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC3713324 Supplier Akina Inc Supplier No. AK141
PLGA-PEG-PLGA (Mw ~1,750-1,500-1,750 Da) LA:GA 15:1 Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)