Learn More
Akina Inc Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-carboxylic acid endcap (per 100mg)
SDP
Catalog No. NC3825722
Encompass
PLGA-PEG-COOH LG 50:50 (Mw ~40,000:5,000 Da) Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-carboxylic acid endcap (per 100mg)
Catalog No. NC3825722 Supplier Akina Inc Supplier No. AI076100MG
PLGA-PEG-COOH LG 50:50 (Mw ~40,000:5,000 Da) Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-carboxylic acid endcap (per 100mg)