Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 1.0 Micron, 25mm, 100/Pk
SDP
Catalog No. NC0641309
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 1 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. NC0641309 Supplier Sterlitech Corporation Supplier No. PET1025100
Sterlitech polyester track etch (PETE) membrane filters, 1 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.