Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methyl methacrylate)

Catalog No. 18223025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-223-025G 25 g
18-223-01KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-223-025G Supplier Sigma Aldrich Supplier No. 18223025G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly (methyl methacrylate) (PMMA) is an amorphous transparent thermoplastic polymer. PMMA is recognized as an optical polymer based on its refractive index of 1.49. Hence, it is used in optical fibers.It finds uses in biological applications because of its lower water absorption capability and biocompatible.The maximum moisture content of PMMA is 1.71% and SBF absorption is 1.96%.Simulated Body fluid test (SBF) is a method to characterize the in vitro bioactivity of ceramic materials, by immersing the materials in an aqueous SBF solution.PMMA/HA (Hydroxyapatite) composites have been used in biomedical applications such as dentistry, orthopedic retainers, and bone replacement. It is used as substrate for graphene growth.Some studies report its use inPMMA-titania hybrid optical thins filmsPMMA/polystryrene/clay nanocompositesPMMA/polyurethane/carbon black nanocomposites for methanol fuel cells

Specifications

CAS 9011-14-7
Density 1.188 g/mL (at 25°C)
Refractive Index n20/D 1.49
Linear Formula [CH2C(CH3)(CO2CH3)]n
MDL Number MFCD00134349
Quantity 25 g
Synonym PMMA; Poly(methacrylic acid methyl ester)
Molecular Weight (g/mol) 120000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.