Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methyl methacrylate)

Catalog No. 445746500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-574-6500G 500 g
44-574-61KG 1 kg
44-574-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 44-574-6500G Supplier Sigma Aldrich Supplier No. 445746500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The thermal degradation behaviour of PMMA ( avg Mol. wt. ∼350,000 g mol-1)was investigated in inert atmosphere, four weight loss steps were observed.Pigment-wetting binder for coatings.

Specifications

CAS 9011-14-7
Density 1.17 g/mL (at 25°C)
Linear Formula [CH2C(CH3)(CO2CH3)]n
MDL Number MFCD00134349
Quantity 500 g
Synonym PMMA; Poly(methacrylic acid methyl ester)
Molecular Weight (g/mol) 350000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.