Learn More
Medchemexpress LLC Poly(methyl methacrylate) | 9011-14-7 | (C5H8O2)n | 500 G
SDP
Catalog No. 5000285190
Encompass_Preferred
Poly(methyl methacrylate) (PMMA) is an amorphous, transparent thermoplastic polymer supplied as solid beads for laboratory and research use. It is commonly used in microfluidic device fabrication, prototyping, and applications that require optical clarity and dimensional stability. The material is provided with supporting documentation for safe handling and analysis.
  • Amorphous thermoplastic with high optical clarity.
  • Available as solid beads for easy handling and dosing.
  • Suitable for microfluidic chip fabrication and prototyping.
  • Resistant to many common solvents and chemicals.
  • Compatible with molding, machining, and lithographic processing.
  • Documentation available: data sheet, SDS, and COA.
Catalog No. 50-002-85190 Supplier Medchemexpress LLC Supplier No. HYW391506500G
Poly(methyl methacrylate) (PMMA) is an amorphous, transparent thermoplastic polymer supplied as solid beads for laboratory and research use. It is commonly used in microfluidic device fabrication, prototyping, and applications that require optical clarity and dimensional stability. The material is provided with supporting documentation for safe handling and analysis.
  • Amorphous thermoplastic with high optical clarity.
  • Available as solid beads for easy handling and dosing.
  • Suitable for microfluidic chip fabrication and prototyping.
  • Resistant to many common solvents and chemicals.
  • Compatible with molding, machining, and lithographic processing.
  • Documentation available: data sheet, SDS, and COA.