Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Poly(methyl methacrylate) | 9011-14-7 | MFCD00134349 | >98.0% | [CH2C(CH3)(CO2CH3)]n | 250 G
SDP

Catalog No. 5000285072
Encompass_Preferred

Poly(methyl methacrylate) (PMMA) is an amorphous thermoplastic polymer provided as solid beads for research applications. It is commonly used as a material for fabrication of microfluidic chips and optical components, and is supplied as a white to off-white solid.

  • Solid beads suitable for molding and microfabrication
  • White to off-white appearance
  • Used for production of microfluidic chips and optical parts
  • Chemical formula: [CH2C(CH3)(CO2CH3)]n; CAS 9011-14-7
  • Storage: 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month)
  • Available in multiple pack sizes: 100 G, 250 G, 500 G, 1 KG

Catalog No. 50-002-85072 Supplier Medchemexpress LLC Supplier No. HYW391506250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(methyl methacrylate) (PMMA) is an amorphous thermoplastic polymer provided as solid beads for research applications. It is commonly used as a material for fabrication of microfluidic chips and optical components, and is supplied as a white to off-white solid.

  • Solid beads suitable for molding and microfabrication
  • White to off-white appearance
  • Used for production of microfluidic chips and optical parts
  • Chemical formula: [CH2C(CH3)(CO2CH3)]n; CAS 9011-14-7
  • Storage: 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month)
  • Available in multiple pack sizes: 100 G, 250 G, 500 G, 1 KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.