Learn More
Medchemexpress LLC Poly(methyl methacrylate) | 9011-14-7 | MFCD00134349 | [CH2C(CH3)(CO2CH3)]n | 1 KG
SDP
Catalog No. 5000285232
Encompass_Preferred
Poly(methyl methacrylate) (PMMA) is a transparent thermoplastic polymer supplied as beads and used for microfabrication applications such as microfluidic chip production. The material provides optical clarity, machinability, and chemical resistance, and is offered in laboratory pack sizes including a 1 kg pack.
  • Good optical clarity for microfluidic device visualization.
  • Machinable and easily patterned for microfabrication processes.
  • Chemical resistance suitable for common laboratory solvents.
  • Available in bead form for molding or solvent casting.
  • Supplied with product documentation such as SDS and COA for safe handling.
Catalog No. 50-002-85232 Supplier Medchemexpress LLC Supplier No. HYW3915061KG
Poly(methyl methacrylate) (PMMA) is a transparent thermoplastic polymer supplied as beads and used for microfabrication applications such as microfluidic chip production. The material provides optical clarity, machinability, and chemical resistance, and is offered in laboratory pack sizes including a 1 kg pack.
  • Good optical clarity for microfluidic device visualization.
  • Machinable and easily patterned for microfabrication processes.
  • Chemical resistance suitable for common laboratory solvents.
  • Available in bead form for molding or solvent casting.
  • Supplied with product documentation such as SDS and COA for safe handling.