Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(methyl methacrylate) for for GPC 4,000, MilliporeSigma™ Supelco™

Catalog No. 111015837 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5837 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5837 Supplier MilliporeSigma Supelco Supplier No. 94131500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 9011-14-7
Ignition Point 580°F
Density 1.200 g/cm3
Flash Point >250°C
Physical Form Neat
Linear Formula [CH2 C(CH3)(CO2 CH3)]n
MDL Number MFCD00134349
Synonym PMMA; Poly(methacrylic acid methyl ester)
Solubility Insoluble in water
Type Analytical Standard
Quantity 500 mg
Grade GPC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.