Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methyl vinyl ether-alt-maleic acid)

Catalog No. 181099100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-109-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-109-9100G Supplier Sigma Aldrich Supplier No. 181099100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Detergent builder.

Specifications

CAS 25153-40-6
Linear Formula [CH2CH(OCH3)CH(CO2H)CH(CO2H)]n
MDL Number MFCD00133997
Quantity 100 g
Synonym Methoxyethylene-maleic acid copolymer; Methyl vinyl ether-maleic acid copolymer; Poly(maleic acid-methyl vinyl ether)
Molecular Weight (g/mol) 1980000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.