Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methyl vinyl ether-alt-maleic acid)

Catalog No. 191124250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-112-4250G 250 g
19-112-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-112-4250G Supplier Sigma Aldrich Supplier No. 191124250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(methyl vinyl ether-alt-maleic acid) (PMVEMA) is a hydrophilic polymer which is biocompatible with bioadhesive properties. It is an acidic polymer that can be used to improve the stability and zeta potential of polyaniline (PANI) solution.PMVEMA can be cross-linked with cellulose whiskers for the formation of nanocomposite films for potential usage in medical and food packaging applcations. It can also be conjugated with polyethylene glycol (PEG) to form hydrogels which can be used in drug delivery systems.

Specifications

CAS 25153-40-6
Linear Formula [CH2CH(OCH3)CH(CO2H)CH(CO2H)]n
MDL Number MFCD00133997
Quantity 250 g
Synonym Maleic acid-methyl vinyl ether copolymer; Methoxyethylene-maleic acid copolymer; Poly(maleic acid-methyl vinyl ether); Vinyl methyl ether-maleic acid copolymer
Molecular Weight (g/mol) 216000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.