Learn More
Medchemexpress LLC Poly(methyl vinyl ether-alt-maleic anhydride) | 9011-16-9 | MFCD00134013 | 98.0% | 216000 g/mol (average) | (C4H2O3·C3H6O)n | 25 G
SDP
Catalog No. 5000285175
Encompass_Preferred
Poly(methyl vinyl ether-alt-maleic anhydride) (average Mw 216,000) is an alternating copolymer used as a polymer reagent to formulate antibacterial hydrogels. It offers a balance of mechanical strength and high swelling capacity, making it suitable for biomaterials research, hydrogel formation, and biochemical assay applications.
  • High average molecular weight provides mechanical strength
  • Excellent swelling ability supports hydrogel networks
  • Suitable for antibacterial hydrogel formulation
  • Stored dry and cool for stability
Catalog No. 50-002-85175 Supplier Medchemexpress LLC Supplier No. HYW39225825G
Poly(methyl vinyl ether-alt-maleic anhydride) (average Mw 216,000) is an alternating copolymer used as a polymer reagent to formulate antibacterial hydrogels. It offers a balance of mechanical strength and high swelling capacity, making it suitable for biomaterials research, hydrogel formation, and biochemical assay applications.
  • High average molecular weight provides mechanical strength
  • Excellent swelling ability supports hydrogel networks
  • Suitable for antibacterial hydrogel formulation
  • Stored dry and cool for stability