Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(-methylstyrene), For GPC, 100,000, MilliporeSigma™ Supelco™

Catalog No. 111013702 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3702 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3702 Supplier MilliporeSigma Supelco Supplier No. 81520250MG
Add to Cart
Add to Cart

Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00152638

Specifications

CAS 25014-31-7
Density 1.075 g/mL (at 25°C)
Linear Formula [CH2 C(CH3)(C6H5)]n
Quantity 250 mg
Grade For GPC
Physical Form Neat
ShelfLife Limited shelf life, expiry date on the label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.