Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly[N-isopropylacrylamide-co-4-(diphenylphosphino)styrene] Palladium(II) Dichloride, TCI America™
SDP

Catalog No. P1425100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1425100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1425100MG Supplier TCI America Supplier No. P1425100MG
Add to Cart
Add to Cart

(ratio, acrylamide:phosphine=20:2)

Chemical Identifiers

MDL Number MFCD04041046
Synonym Palladium Catalyst, supported with Poly[N-isopropylacrylamide-co-4-(diphenylphosphino)styrene]

Specifications

Color Black
Quantity 100 mg
Physical Form Crystalline Powder
Chemical Name or Material Poly[N-isopropylacrylamide-co-4-(diphenylphosphino)styrene] Palladium(II) Dichloride (ratio, acrylam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.