Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(neopentyl Glycol Succinate), TCI America™
SDP

Catalog No. S00995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00995G Supplier TCI America Supplier No. S00995G

Chemical Identifiers

CAS 28257-92-3
Molecular Formula C9H18O6
Molecular Weight (g/mol) 222.237
MDL Number MFCD00148032
InChI Key GXTJJXHLTYJRLX-UHFFFAOYSA-N
Synonym Neopentyl Glycol Succinate Polyester, Succinate Neopentylglycol Polyester
PubChem CID 57376614
IUPAC Name butanedioic acid;2,2-dimethylpropane-1,3-diol
SMILES CC(C)(CO)CO.C(CC(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Poly(neopentyl Glycol Succinate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.