Learn More
MSC Akro-Mils Nest and Stack Storage Tote, Polypropylene, 19.4 Gal., 3/Pk.
Catalog No. 5000516473
Encompass
This Nest and Stack storage tote is designed for efficient inventory control and organized storage in industrial environments. Engineered for versatility, these bins stack securely when full and can be turned 180 degrees to nest compactly when empty, helping to save space. The durable, one-piece construction is well-suited for heavy-duty applications, providing resistance to water, rust, corrosion, and weak acids.
  • Storage capacity of 19.4 gal.
  • Load capacity of 90 lb. and a stacking capacity of 450 lb.
  • Overall dimensions of 23-1/2 in. L x 19-1/2 in. W x 13 in. H
  • Industrial-grade polymer construction
  • Compatible with optional lids and label holders
  • Space-saving nesting and stacking design
Catalog No. 50-005-16473 Supplier MSC Supplier No. 43398791
This Nest and Stack storage tote is designed for efficient inventory control and organized storage in industrial environments. Engineered for versatility, these bins stack securely when full and can be turned 180 degrees to nest compactly when empty, helping to save space. The durable, one-piece construction is well-suited for heavy-duty applications, providing resistance to water, rust, corrosion, and weak acids.
  • Storage capacity of 19.4 gal.
  • Load capacity of 90 lb. and a stacking capacity of 450 lb.
  • Overall dimensions of 23-1/2 in. L x 19-1/2 in. W x 13 in. H
  • Industrial-grade polymer construction
  • Compatible with optional lids and label holders
  • Space-saving nesting and stacking design