Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(octadecyl methacrylate) solution

Catalog No. 18233825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-233-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-233-825G Supplier Sigma Aldrich Supplier No. 18233825G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(octadecyl methacrylate) may be used to prepare block copolymers to form molecular brushes.Unit weight does not include weight of solvent.

Specifications

CAS 25639-21-8
Linear Formula (C22H42O2)n
Quantity 25 g
MDL Number MFCD00084444
Synonym Poly(octadecyl methacrylate); Poly(stearyl methacrylate)
Molecular Weight (g/mol) 170000
Percent Purity 30-35 wt.% (in toluene)
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.