Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-(2-aminoethyl)-ω-(carboxymethoxy)- | 139729-28-5 | 1100 (Average) | 25 MG
SDP
Catalog No. 5000394757
Encompass_Preferred
NH2-PEG-COOH (Mn 1100) is a bifunctional cross-linker intended for research use only. It can be used to synthesize PEG hydrogels.
  • Bifunctional cross-linker
  • Used to synthesize PEG hydrogels
  • White to off-white solid appearance
Catalog No. 50-003-94757 Supplier Medchemexpress LLC Supplier No. 25MGHY168940
NH2-PEG-COOH (Mn 1100) is a bifunctional cross-linker intended for research use only. It can be used to synthesize PEG hydrogels.
  • Bifunctional cross-linker
  • Used to synthesize PEG hydrogels
  • White to off-white solid appearance