Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-(2-aminoethyl)-ω-methoxy- | 80506-64-5 | 98.0% | (C2H4O)nC3H9NO | 250 MG
SDP
Catalog No. 5000352758
Encompass_Preferred
mPEG-amine (MW 5000) is a modifier that increases the water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes by replacing the sulfonic acid portion of the dye molecule. It can also form amide bonds with carboxyl groups on the surface of microspheres through EDC and Sulfo-NHS mediation, creating a PEG coating on fluorescent microspheres for large-scale rotational cytoplasmic flow studies. This product is for research use only.
  • Increases water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes
  • Replaces the sulfonic acid portion of dye molecules
  • Forms amide bonds with carboxyl groups on microsphere surfaces
  • Creates a PEG coating on fluorescent microspheres
  • Suitable for large-scale rotational cytoplasmic flow studies
  • For research use only
Catalog No. 50-003-52758 Supplier Medchemexpress LLC Supplier No. 250MGHY140677
mPEG-amine (MW 5000) is a modifier that increases the water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes by replacing the sulfonic acid portion of the dye molecule. It can also form amide bonds with carboxyl groups on the surface of microspheres through EDC and Sulfo-NHS mediation, creating a PEG coating on fluorescent microspheres for large-scale rotational cytoplasmic flow studies. This product is for research use only.
  • Increases water solubility of long-wavelength voltage-sensitive dyes (VSD) or Pittsburgh (PGH) dyes
  • Replaces the sulfonic acid portion of dye molecules
  • Forms amide bonds with carboxyl groups on microsphere surfaces
  • Creates a PEG coating on fluorescent microspheres
  • Suitable for large-scale rotational cytoplasmic flow studies
  • For research use only