Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-(2-aminoethyl)-ω-methoxy- | 80506-64-5 | 98.0% | (C2H4O)nC3H9NO | 50 MG
SDP
Catalog No. 5000354937
Encompass_Preferred
mPEG-amine (MW 20000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These compounds exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54937 Supplier Medchemexpress LLC Supplier No. HY14067950MG
mPEG-amine (MW 20000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. These compounds exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins