Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-[6-[[(3β)-cholest-5-en-3-yl]oxy]-1,6-dioxohexyl]-ω-[[6-[[(3β)-cho | 69068-97-9 | MFCD00151772 | 1055.60 g/mol | C68H110O8 | 100 MG
SDP
Catalog No. 5000283629
Encompass_Preferred
Cholesterol-PEG 600 is a cholesterol-polyethylene glycol conjugate used as an amphiphilic lipid modifier for liposome formulation and related life-science research. It enhances steric stabilization and aqueous compatibility of lipid assemblies, and is supplied as an off-white to light yellow oil with documented storage recommendations and batch-level quality documentation.
Catalog No. 50-002-83629 Supplier Medchemexpress LLC Supplier No. HYW127558100MG
Cholesterol-PEG 600 is a cholesterol-polyethylene glycol conjugate used as an amphiphilic lipid modifier for liposome formulation and related life-science research. It enhances steric stabilization and aqueous compatibility of lipid assemblies, and is supplied as an off-white to light yellow oil with documented storage recommendations and batch-level quality documentation.