Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-[(9R)-6-hydroxy-6-oxido-1,12-dioxo-9-[(1-oxooctadecyl)oxy]-5,7,11-trioxa-2-aza-6-phosphanonacos-1-yl]-ω-[2-[(3-carboxy-1-oxopropyl)amino]ethoxy]- | 1445832-74-5 | 99.95% | 2000 (Average) | 5 MG
SDP
Catalog No. 5000384511
Encompass_Preferred
DSPE-PEG (2000)-COOH is a pegylated derivative of 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine. It can form micelles for targeted drug delivery.
  • Can form micelles for targeted drug delivery
  • Appearance: solid
  • Color: white to off-white
Catalog No. 50-003-84511 Supplier Medchemexpress LLC Supplier No. HY1707065MG
DSPE-PEG (2000)-COOH is a pegylated derivative of 1,2-Distearoyl-sn-glycero-3-phosphorylethanolamine. It can form micelles for targeted drug delivery.
  • Can form micelles for targeted drug delivery
  • Appearance: solid
  • Color: white to off-white