Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-hydro-ω-hydroxy | 25322-68-3 | 1,000,000 (average) | (C2H4O)nH2O | 100g
SDP
Catalog No. 5000268422
Encompass_Preferred
PEG1000000 (Polyethylene glycol 1000000) is a polymer compound with good biocompatibility and biodegradability and is widely used in drug controlled release systems and bioadhesive materials[1]
Catalog No. 50-002-68422 Supplier Medchemexpress LLC Supplier No. HYY0873A3100G
PEG1000000 (Polyethylene glycol 1000000) is a polymer compound with good biocompatibility and biodegradability and is widely used in drug controlled release systems and bioadhesive materials[1]