Learn More
Medchemexpress LLC Poly(oxy-1,2-ethanediyl), α-hydro-ω-hydroxy- | 25322-68-3 | MFCD00081839 | 500.00 g/mol | (C2H4O)nH2O | 100g
SDP
Catalog No. 5000268424
Encompass_Preferred
PEG500 (Polyethylene glycol 500) is a versatile polymer that s used as a solvent dispersant lubricant and more in industries like pharmaceuticals and biotechnology and it can also modify liposomes PEG500 is commonly used as a solvent or carrier for drugs helping with capsule endoscopy to improve image quality and cecum completion rates[1][2]
Catalog No. 50-002-68424 Supplier Medchemexpress LLC Supplier No. HYY0873M100G
PEG500 (Polyethylene glycol 500) is a versatile polymer that s used as a solvent dispersant lubricant and more in industries like pharmaceuticals and biotechnology and it can also modify liposomes PEG500 is commonly used as a solvent or carrier for drugs helping with capsule endoscopy to improve image quality and cecum completion rates[1][2]