Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly[(p-phenylenevinylene)-alt-(2-methoxy-5-(2-ethylhexyloxy)-p-phenylenevinylene)]

Catalog No. 5941991G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-419-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-419-91G Supplier Sigma Aldrich Supplier No. 5941991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly[(p-phenylenevinylene)-alt-(2-methoxy-5-(2-ethylhexyloxy)-p-phenylenevinylene)] (p-PMEH-PPV) is a conducting polymer that can be used as an active material in the development of optoelectronic devices. It forms a conjugated-nonconjugated multiblock copolymer which can be used as an electroluminescent polymeric material. It can be potentially operated at low voltages with quick responsive and defined displays.p-PMEH-PPV can be used as a blue EL polymer for the fabrication of semiconductor devices such as organic light emitting diodes (OLEDs), thin film transistors (TFTs), photovoltaic cells and chemical sensors.

Specifications

CAS 211678-36-3
Linear Formula (C25H32O2)n
Quantity 1 g
MDL Number MFCD05665553
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.