Learn More
MSC PIP Cut Resistant Gloves, Size XL, A2 Cut Level, Nitrile Coated, 100/Cs.
Catalog No. 5000508806
Encompass
These high-performance cut-resistant gloves provide superior protection for demanding industrial applications. Engineered with a specialized 18-gauge knit liner, the gloves offer a balance of dexterity, tactile sensitivity, and cut protection. They feature a foam nitrile palm coating that enhances grip in dry, damp, and oily conditions, while the integrated knit wrist ensures a secure fit.
  • Cut level A2 protection
  • Foam nitrile palm and finger coating for improved grip
  • Reinforced thumb crotch for increased durability in high-wear areas
  • Seamless knit construction with recycled polyester, PE steel fiber, and spandex
  • ESD compliant and touchscreen compatible
  • Food certified per FDA 21CFR177.2600 and EC 1935/2004
  • Fiberglass-free composition
  • Reusable and launderable at 40°C
Catalog No. 50-005-08806 Supplier MSC Supplier No. 37097615
These high-performance cut-resistant gloves provide superior protection for demanding industrial applications. Engineered with a specialized 18-gauge knit liner, the gloves offer a balance of dexterity, tactile sensitivity, and cut protection. They feature a foam nitrile palm coating that enhances grip in dry, damp, and oily conditions, while the integrated knit wrist ensures a secure fit.
  • Cut level A2 protection
  • Foam nitrile palm and finger coating for improved grip
  • Reinforced thumb crotch for increased durability in high-wear areas
  • Seamless knit construction with recycled polyester, PE steel fiber, and spandex
  • ESD compliant and touchscreen compatible
  • Food certified per FDA 21CFR177.2600 and EC 1935/2004
  • Fiberglass-free composition
  • Reusable and launderable at 40°C