Learn More
Uline POLY PEGBOARD - 48 X 24"",BLACK
Catalog No. NC1957161
Encompass
Will not rust, crack or break.Recommended cleanrooms and repair areas.Commercial-grade 1/4" polypropylene.9/32" diameter holes are 1" apart.
Catalog No. NC1957161 Supplier Uline Supplier No. H7791BL
Will not rust, crack or break.Recommended cleanrooms and repair areas.Commercial-grade 1/4" polypropylene.9/32" diameter holes are 1" apart.