Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(pentafluorostyrene)

Catalog No. 591947200MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-194-7200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-194-7200MG Supplier Sigma Aldrich Supplier No. 591947200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(pentafluorostyrene) (PPFS) can be used in the development of polylectrolytic membranes for fuel cell applications. It can also act as a glycosylated polymer for potential usage in biological imaging. It is a fluorinated polymer which may be utilized in microelectronics or as an antifouling and antifogging agent.

Specifications

CAS 26838-55-1
Linear Formula (C8H3F5)n
MDL Number MFCD03458138
Quantity 200 mg
Synonym 2,3,4,5,6-Pentafluorostyrene homopolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.