Learn More
MSC Carlisle Kettle Brush, 5-1/2 x 9-1/2 in, Polyester, 1 ea
Catalog No. 502867505
Encompass
This kettle brush is designed for use in foodservice, commercial kitchens, and food processing equipment. It is also color-coded for easy identification.
  • Brush type: Food service
  • Bristle material: Polyester
  • Block material: Plastic
  • Brush length: 9-1/2 in
  • Brush width: 5-1/2 in
  • Bristle firmness: Stiff
  • Bristle color: Blue
  • Resistance features: Water-resistant; Oil-resistant
Catalog No. 50-286-7505 Supplier MSC Supplier No. 19601681
This kettle brush is designed for use in foodservice, commercial kitchens, and food processing equipment. It is also color-coded for easy identification.
  • Brush type: Food service
  • Bristle material: Polyester
  • Block material: Plastic
  • Brush length: 9-1/2 in
  • Brush width: 5-1/2 in
  • Bristle firmness: Stiff
  • Bristle color: Blue
  • Resistance features: Water-resistant; Oil-resistant