Learn More
MSC Carlisle Hand Scrub, Color Coded, 6 in, White, 1 ea
Catalog No. 502816353
Encompass
This hand scrub brush is designed for commercial establishments, featuring a color-coded system to support sanitary maintenance protocols. It is constructed from durable materials to withstand rigorous use.
  • Polyester bristles for effective scrubbing
  • Plastic block material for durability
  • Medium bristle firmness suitable for various cleaning tasks
  • Resistant to water and oil for extended use
  • FDA compliant for food service and hygienic applications
  • Color-coded for easy identification within sanitary maintenance systems
Catalog No. 50-281-6353 Supplier MSC Supplier No. 10785376
This hand scrub brush is designed for commercial establishments, featuring a color-coded system to support sanitary maintenance protocols. It is constructed from durable materials to withstand rigorous use.
  • Polyester bristles for effective scrubbing
  • Plastic block material for durability
  • Medium bristle firmness suitable for various cleaning tasks
  • Resistant to water and oil for extended use
  • FDA compliant for food service and hygienic applications
  • Color-coded for easy identification within sanitary maintenance systems