Learn More
MSC AbilityOne Plastic Sheeting, 100 x 200 ft, 1 Roll
Catalog No. 502869403
Encompass
This polyethylene plastic sheeting is designed for general-purpose packaging, surface protection, construction, and agricultural uses. It provides protection for surfaces from dust and is both highly water-resistant and moderately vapor-resistant.
  • Highly water-resistant
  • Moderately vapor-resistant
  • Protects surfaces from dust
  • Suitable for general-purpose packaging, surface protection, construction, and agricultural applications
  • Made to LP-378 standards
Catalog No. 50-286-9403 Supplier MSC Supplier No. 53386017
This polyethylene plastic sheeting is designed for general-purpose packaging, surface protection, construction, and agricultural uses. It provides protection for surfaces from dust and is both highly water-resistant and moderately vapor-resistant.
  • Highly water-resistant
  • Moderately vapor-resistant
  • Protects surfaces from dust
  • Suitable for general-purpose packaging, surface protection, construction, and agricultural applications
  • Made to LP-378 standards