Learn More
MSC Carlisle Valve & Fitting Brush, Polyester, 30 in Long, 3 in Diameter
Catalog No. 502839859
Encompass
This multi-purpose brush is designed for foodservice, commercial kitchens, and food processing equipment. Featuring durable polyester bristles, it measures 30 inches in overall length with a 3-inch bristle diameter, making it suitable for a variety of cleaning tasks.
  • Versatile for cleaning commercial kitchen equipment
  • Color-coded design helps reduce cross-contamination
  • Rounded head and ends protect against scratching surfaces
  • Cleans effectively in all directions
  • Medium stiff polyester bristles provide aggressive cleaning
  • BPA free and made with FDA compliant block and bristles
  • Convenient hanging hole on handle for easy storage
Catalog No. 50-283-9859 Supplier MSC Supplier No. 95753877
This multi-purpose brush is designed for foodservice, commercial kitchens, and food processing equipment. Featuring durable polyester bristles, it measures 30 inches in overall length with a 3-inch bristle diameter, making it suitable for a variety of cleaning tasks.
  • Versatile for cleaning commercial kitchen equipment
  • Color-coded design helps reduce cross-contamination
  • Rounded head and ends protect against scratching surfaces
  • Cleans effectively in all directions
  • Medium stiff polyester bristles provide aggressive cleaning
  • BPA free and made with FDA compliant block and bristles
  • Convenient hanging hole on handle for easy storage