Learn More
MSC Carlisle Multi-Purpose Valve & Fitting Brush, Polyester, 30 in, 4 in Diameter
Catalog No. 502840045
Encompass
The multi-purpose valve and fitting brush is designed for versatile and aggressive cleaning of commercial kitchen equipment. It features durable polyester bristles and a rounded head to protect surfaces from scratching during use.
  • Round or oval head is versatile for cleaning a variety of commercial kitchen equipment.
  • Rounded heads and ends protect against scratching.
  • Cleans effectively in all directions.
  • Medium stiff polyester bristles provide aggressive cleaning.
  • BPA free materials.
  • Made with FDA compliant block and bristles.
Catalog No. 50-284-0045 Supplier MSC Supplier No. 95753901
The multi-purpose valve and fitting brush is designed for versatile and aggressive cleaning of commercial kitchen equipment. It features durable polyester bristles and a rounded head to protect surfaces from scratching during use.
  • Round or oval head is versatile for cleaning a variety of commercial kitchen equipment.
  • Rounded heads and ends protect against scratching.
  • Cleans effectively in all directions.
  • Medium stiff polyester bristles provide aggressive cleaning.
  • BPA free materials.
  • Made with FDA compliant block and bristles.