Learn More
Bio Rad Laboratories POLY-PREP CHROMATOGRAPHY COLUM
Supplier:  Bio Rad Laboratories 7311553
Encompass
Column, Chromatography Bio-Rad Poly-prep Conical Empty Includes end caps, tip closures, 1,000 Autoclavable Holds upto 2mL of chromatography support and 10mL of eluent or sample in an integral reservoir 9cm high 0.8 x 4cm PP column
Catalog No. 50-901-1598