Learn More
Bio Rad Laboratories POLY-PREP COLUMNS
Catalog No. 509011605
Encompass
Column Poly-Prep Bio-Rad AG 50W-X8 resin 200 to 400 mesh PP w/end cap 10mL reservoir 2mL bed volume Graduated Porous 20um PE bed support Snap-off tip 63-150um particle Ionic form hydrogen
Catalog No. 50-901-1605 Supplier Bio Rad Laboratories Supplier No. 7316214
Column Poly-Prep Bio-Rad AG 50W-X8 resin 200 to 400 mesh PP w/end cap 10mL reservoir 2mL bed volume Graduated Porous 20um PE bed support Snap-off tip 63-150um particle Ionic form hydrogen