Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(propylene glycol) bis(2-aminopropyl ether)

Catalog No. 406694250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-669-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-669-4250ML Supplier Sigma Aldrich Supplier No. 406694250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(propylene glycol) bis(2-aminopropyl ether)

Specifications

CAS 9046-10-0
Density 0.997 g/mL (at 25°C)
Refractive Index n20/D 1.452
Linear Formula CH3CH(NH2)CH2[OCH2CH(CH3)]nNH2
MDL Number MFCD00082259
Quantity 250 mL
Molecular Weight (g/mol) 4000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.