Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(propylene glycol) monobutyl ether

Catalog No. 4381111L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-811-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-811-11L Supplier Sigma Aldrich Supplier No. 4381111L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydraulic fluids, metal working fluids and lubricants, heat transfer fluids, solder assist fluids, quenchants, lubricants, solvents, plasticizers and foam control agents.

Specifications

CAS 9003-13-8
Density 0.989 g/mL (at 25°C)
Boiling Point >200°C
Linear Formula CH3(CH2)3 [OCH2CH(CH3)]nOH
Refractive Index n20/D 1.448
MDL Number MFCD00148010
Quantity 1 L
Synonym Butoxypolypropylene glycol; PPG-14 Butyl Ether; PPG-16 Butyl Ether; Poly(propylene oxide) monobutyl ether; Polyoxypropylene glycol butyl monoether; Polypropylene glycol butyl ether; Polypropylene glycol monobutyl ether
Molecular Weight (g/mol) 1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.