Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(propylene glycol), tolylene 2,4-diisocyanate terminated

Catalog No. 433497250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-349-7250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-349-7250ML Supplier Sigma Aldrich Supplier No. 433497250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Idler rollers and adhesive-handling pads.

Specifications

CAS 9057-91-4
Density 1.05 g/mL (at 25°C)
Refractive Index n20/D 1.4742
MDL Number MFCD00217728
Quantity 250 mL
Synonym Polypropylene glycol-toluene diisocyanate copolymer; Tolylene diisocyanate-polypropylene glycol polymer
Molecular Weight (g/mol) 2300
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.