Learn More
Vector Laboratories Polyclonal Rabbit anti‑Human MSH3 Antibody (IF, WB) 100ul
SDP
Catalog No. NC3595136
Encompass
MSH3 antibody LS-C804049 is an unconjugated rabbit polyclonal antibody to MSH3 from human. It is reactive with human, mouse and rat. Validated for IF, Peptide-ELISA and WB.
Catalog No. NC3595136 Supplier Vector Laboratories Supplier No. LSC804049100
MSH3 antibody LS-C804049 is an unconjugated rabbit polyclonal antibody to MSH3 from human. It is reactive with human, mouse and rat. Validated for IF, Peptide-ELISA and WB.