Learn More
Vector Laboratories POLY RAIV H9 HEMAGG ANTI 100UL
SDP
Supplier:  Vector Laboratories LSC486880100
Encompass
Antibody: Influenza A virus H9 Hemagglutinin Rabbit anti-influenza virus Polyclonal Antibody.Application: IHC-P, ICC,IF,WB,IP,Flo, ELISA.Reactivity: Influenza VirusFormat:Unconjugated,Unmodified100ul, LIFESPAN BIOSCIENCES mfg# LS-C486880-100
Catalog No. NC2078086